Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-[3-(aminomethyl)phenyl]propionate, 99%

$124.68 - $434.08

Chemical Identifiers

Chemical Name or Material Methyl 3-[3-(aminomethyl)phenyl]propionate
CAS 100511-83-9
Molecular Formula C11H15NO2
Formula Weight 193.25
MDL Number MFCD08064274
Synonym 3-[3-(Aminomethyl)phenyl]propionic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5094103

 
alfa aesar™
H5094103
1g Each for $434.08

AAH50941MD

 
alfa aesar™
H50941MD
250mg Each for $124.68
Description & Specifications

Specifications

Formula Weight 193.25
Chemical Name or Material Methyl 3-[3-(aminomethyl)phenyl]propionate
Assay 99%
Molecular Formula C11H15NO2
MDL Number MFCD08064274
CAS 100511-83-9
Synonym 3-[3-(Aminomethyl)phenyl]propionic acid methyl ester
Quantity 1g

Applications
Methyl 3-[3-(aminomethyl)phenyl]propionate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.