missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 3-(2-morpholin-4-ylethoxy)benzoate, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 249937-00-6 |
---|---|
Molecular Formula | C14H19NO4 |
Molecular Weight (g/mol) | 265.31 |
MDL Number | MFCD08060518 |
InChI Key | VOYSHDFODOJUMP-UHFFFAOYSA-N |
Synonym | methyl 3-2-morpholin-4-ylethoxy benzoate, methyl 3-2-morpholin-4-yl ethoxy benzoate, methyl 3-2-morpholinoethoxy benzoate, methyl 3-2-morpholin4-ylethoxy benzoate, 3-2-morpholin-4-yl-ethoxy-benzoic acid methyl ester, benzoic acid,3-2-4-morpholinyl ethoxy-, methyl ester |
PubChem CID | 7164628 |
IUPAC Name | methyl 3-(2-morpholin-4-ylethoxy)benzoate |
SMILES | COC(=O)C1=CC(OCCN2CCOCC2)=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC42022DA
|
Thermo Scientific Chemicals
CC42022DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC42022DE
|
Thermo Scientific Chemicals
CC42022DE |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
249937-00-6 | |
265.31 | |
VOYSHDFODOJUMP-UHFFFAOYSA-N | |
7164628 | |
COC(=O)C1=CC(OCCN2CCOCC2)=CC=C1 |
C14H19NO4 | |
MFCD08060518 | |
methyl 3-2-morpholin-4-ylethoxy benzoate, methyl 3-2-morpholin-4-yl ethoxy benzoate, methyl 3-2-morpholinoethoxy benzoate, methyl 3-2-morpholin4-ylethoxy benzoate, 3-2-morpholin-4-yl-ethoxy-benzoic acid methyl ester, benzoic acid,3-2-4-morpholinyl ethoxy-, methyl ester | |
methyl 3-(2-morpholin-4-ylethoxy)benzoate |
Specifications
249937-00-6 | |
100.0 | |
Amber glass bottle | |
MFCD08060518 | |
methyl 3-2-morpholin-4-ylethoxy benzoate, methyl 3-2-morpholin-4-yl ethoxy benzoate, methyl 3-2-morpholinoethoxy benzoate, methyl 3-2-morpholin4-ylethoxy benzoate, 3-2-morpholin-4-yl-ethoxy-benzoic acid methyl ester, benzoic acid,3-2-4-morpholinyl ethoxy-, methyl ester | |
COC(=O)C1=CC(OCCN2CCOCC2)=CC=C1 | |
265.31 | |
265.31 | |
Methyl 3-(2-morpholin-4-ylethoxy)benzoate |
96.5 | |
97% | |
C14H19NO4 | |
1g | |
VOYSHDFODOJUMP-UHFFFAOYSA-N | |
methyl 3-(2-morpholin-4-ylethoxy)benzoate | |
7164628 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.