missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 3-(2-hydroxyphenyl)propionate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1221914

 View more versions of this product

Catalog No. AAA1221914



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 3- (2-hydroxyphenyl)propionate
40°C to 44°C
MFCD00067757
methyl 3-2-hydroxyphenyl propanoate, methyl 3-2-hydroxyphenyl propionate, methyl melilotate, acmc-20aki4, methyl hydroxy-benzenepropionate, methyl 2'-hydroxyhydrocinnamate, methyl 3-hydroxyphenyl propionate, methyl 3 2-hydroxyphenyl propanoate, 2-hydroxybenzenepropanoic acid methyl ester, 3-2-hydroxyphenyl propionic acid methyl ester
COC(=O)CCC1=CC=CC=C1O
180.203
180.21
20349-89-7
C10H12O3
25 g
YHXYRISRGHSPNV-UHFFFAOYSA-N
methyl 3-(2-hydroxyphenyl)propanoate
2794569
97%

Chemical Identifiers

20349-89-7
180.203
YHXYRISRGHSPNV-UHFFFAOYSA-N
2794569
COC(=O)CCC1=CC=CC=C1O
C10H12O3
MFCD00067757
methyl 3-2-hydroxyphenyl propanoate, methyl 3-2-hydroxyphenyl propionate, methyl melilotate, acmc-20aki4, methyl hydroxy-benzenepropionate, methyl 2'-hydroxyhydrocinnamate, methyl 3-hydroxyphenyl propionate, methyl 3 2-hydroxyphenyl propanoate, 2-hydroxybenzenepropanoic acid methyl ester, 3-2-hydroxyphenyl propionic acid methyl ester
methyl 3-(2-hydroxyphenyl)propanoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only