Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 3-(1-pyrrolyl)thiophene-2-carboxylate, 97%

$246.28 - $246.28

Chemical Identifiers

Chemical Name or Material Methyl 3-(1-pyrrolyl)thiophene-2-carboxylate
CAS 74772-16-0
Molecular Formula C10H9NO2S
Formula Weight 207.25
MDL Number MFCD00053052
Beilstein 4679860
Synonym 3-(1-Pyrrolyl)thiophene-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0936806

 
alfa aesar™
L0936806
5g Each for $246.28
Description & Specifications

Specifications

Formula Weight 207.25
Chemical Name or Material Methyl 3-(1-pyrrolyl)thiophene-2-carboxylate
Assay 97%
Molecular Formula C10H9NO2S
CAS 74772-16-0
Beilstein 4679860
Synonym 3-(1-Pyrrolyl)thiophene-2-carboxylic acid methyl ester
Quantity 5g
MDL Number MFCD00053052
Melting Point 59° to 63°C