Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate, 98%

$117.61 - $1,268.36

Chemical Identifiers

Chemical Name or Material Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate
CAS 200573-11-1
Molecular Formula C13H9F3O5S
Formula Weight 334.27
MDL Number MFCD04039773
Synonym Methyl 2-trifluoromethanesulfonyloxy-1-naphthoate; 2-Trifluoromethylsulfonyloxy-1-naphthoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2479306

 
alfa aesar™
B2479306
5g Each for $117.61

AAB2479314

 
alfa aesar™
B2479314
25g Each for $415.61

AAB2479322

 
alfa aesar™
B2479322
100g Each for $1,268.36
Description & Specifications

Specifications

Formula Weight 334.27
Chemical Name or Material Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate
Assay 98%
Molecular Formula C13H9F3O5S
MDL Number MFCD04039773
CAS 200573-11-1
Synonym Methyl 2-trifluoromethanesulfonyloxy-1-naphthoate; 2-Trifluoromethylsulfonyloxy-1-naphthoic acid methyl ester
Health Hazard 1 Warning
Melting Point 41° to 43°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.