missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-(Trifluoromethyl)benzoate, TCI America™

$48.89 - $114.58

Chemical Identifiers

CAS 344-96-7
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00236050
InChI Key JLCAMDSWNZBTEE-UHFFFAOYSA-N
Synonym methyl 2-trifluoromethyl benzoate, benzoic acid, trifluoromethyl-, methyl ester, acmc-1ahfp, methyl o-trifluoromethylbenzoate, jlcamdswnzbtee-uhfffaoysa, methyl-2-trifluoromethyl benzoate, 2-trifluoromethylbenzoic acid methyl ester, 2-trifluoromethyl-benzoic acid methyl ester, 2-trifluoromethyl benzoic acid methyl ester, methyl alpha,alpha,alpha-trifluoro-o-toluate
PubChem CID 2775578
IUPAC Name methyl 2-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC=CC=C1C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24755G View Documents TCI America
M24755G
5g Each for $48.89
 
M247525G View Documents TCI America
M247525G
25g Each for $114.58
 
Specifications

Chemical Identifiers

344-96-7
204.148
JLCAMDSWNZBTEE-UHFFFAOYSA-N
2775578
COC(=O)C1=CC=CC=C1C(F)(F)F
C9H7F3O2
MFCD00236050
methyl 2-trifluoromethyl benzoate, benzoic acid, trifluoromethyl-, methyl ester, acmc-1ahfp, methyl o-trifluoromethylbenzoate, jlcamdswnzbtee-uhfffaoysa, methyl-2-trifluoromethyl benzoate, 2-trifluoromethylbenzoic acid methyl ester, 2-trifluoromethyl-benzoic acid methyl ester, 2-trifluoromethyl benzoic acid methyl ester, methyl alpha,alpha,alpha-trifluoro-o-toluate
methyl 2-(trifluoromethyl)benzoate

Specifications

105°C
5g
C9H7F3O2
methyl 2-trifluoromethyl benzoate, benzoic acid, trifluoromethyl-, methyl ester, acmc-1ahfp, methyl o-trifluoromethylbenzoate, jlcamdswnzbtee-uhfffaoysa, methyl-2-trifluoromethyl benzoate, 2-trifluoromethylbenzoic acid methyl ester, 2-trifluoromethyl-benzoic acid methyl ester, 2-trifluoromethyl benzoic acid methyl ester, methyl alpha,alpha,alpha-trifluoro-o-toluate
COC(=O)C1=CC=CC=C1C(F)(F)F
204.148
204.15
≥98.0% (GC)
Colorless
344-96-7
MFCD00236050
JLCAMDSWNZBTEE-UHFFFAOYSA-N
methyl 2-(trifluoromethyl)benzoate
2775578
Clear Liquid at 20°C
Methyl 2-(Trifluoromethyl)benzoate
Safety and Handling

Safety and Handling

TSCA : No