Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-(trifluoromethyl)benzoate, 97%

$58.81 - $229.35

Chemical Identifiers

Chemical Name or Material Methyl 2-(trifluoromethyl)benzoate
CAS 344-96-7
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD00236050
Beilstein 2580873
Synonym 2-(Trifluoromethyl)benzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1413203

  SDS
alfa aesar™
L1413203
1g Each for $58.81

AAL1413206

  SDS
alfa aesar™
L1413206
5g Each for $229.35
Description & Specifications

Specifications

Formula Weight 204.15
Chemical Name or Material Methyl 2-(trifluoromethyl)benzoate
Assay 97%
Molecular Formula C9H7F3O2
CAS 344-96-7
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
MDL Number MFCD00236050
Beilstein 2580873
Synonym 2-(Trifluoromethyl)benzoic acid methyl ester
Health Hazard 1 Warning
Boiling Point 104° to 105°C (25mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.