Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-thiopheneacetate, 98%

$64.34 - $64.34

Chemical Identifiers

Chemical Name or Material Methyl 2-thiopheneacetate
CAS 19432-68-9
Molecular Formula C7H8O2S
Formula Weight 156.2
MDL Number MFCD00041213
Beilstein 120622
Synonym Methyl 2-thienylacetate; 2-Thiopheneacetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1211006

  SDS
alfa aesar™
A1211006
5g Each for $64.34
Description & Specifications

Specifications

Formula Weight 156.2
Chemical Name or Material Methyl 2-thiopheneacetate
Assay 98%
Molecular Formula C7H8O2S
MDL Number MFCD00041213
CAS 19432-68-9
Beilstein 120622
Synonym Methyl 2-thienylacetate; 2-Thiopheneacetic acid methyl ester
Boiling Point 100° to 104°C (14mmHg)
Quantity 5g