missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Pyrazinecarboxylate, TCI America™

$100.83 - $302.50

Chemical Identifiers

CAS 6164-79-0
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00014611
InChI Key TWIIRMSFZNYMQE-UHFFFAOYSA-N
Synonym methyl 2-pyrazinecarboxylate, methyl pyrazinoate, pyrazinoic acid methyl ester, methylpyrazine-2-carboxylate, pyrazinecarboxylic acid, methyl ester, methyl pyrazinecarboxylate, 2-pyrazinecarboxylic acid, methyl ester, pyrazine-2-carboxylic acid, methyl ester, 2-methylpyrazine carboxylate, pyrazine-2-carboxylic acid methyl ester
PubChem CID 72662
IUPAC Name methyl pyrazine-2-carboxylate
SMILES COC(=O)C1=NC=CN=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M201525G View Documents TCI America
M201525G
25g Each for $302.50
 
M20155G View Documents TCI America
M20155G
5g Each for $100.83
 
Specifications

Chemical Identifiers

6164-79-0
138.126
TWIIRMSFZNYMQE-UHFFFAOYSA-N
72662
COC(=O)C1=NC=CN=C1
C6H6N2O2
MFCD00014611
methyl 2-pyrazinecarboxylate, methyl pyrazinoate, pyrazinoic acid methyl ester, methylpyrazine-2-carboxylate, pyrazinecarboxylic acid, methyl ester, methyl pyrazinecarboxylate, 2-pyrazinecarboxylic acid, methyl ester, pyrazine-2-carboxylic acid, methyl ester, 2-methylpyrazine carboxylate, pyrazine-2-carboxylic acid methyl ester
methyl pyrazine-2-carboxylate

Specifications

104°C
60°C
6164-79-0
MFCD00014611
TWIIRMSFZNYMQE-UHFFFAOYSA-N
methyl pyrazine-2-carboxylate
72662
Crystal-Powder at 20°C
Methyl 2-Pyrazinecarboxylate
White-Yellow
25g
C6H6N2O2
methyl 2-pyrazinecarboxylate, methyl pyrazinoate, pyrazinoic acid methyl ester, methylpyrazine-2-carboxylate, pyrazinecarboxylic acid, methyl ester, methyl pyrazinecarboxylate, 2-pyrazinecarboxylic acid, methyl ester, pyrazine-2-carboxylic acid, methyl ester, 2-methylpyrazine carboxylate, pyrazine-2-carboxylic acid methyl ester
COC(=O)C1=NC=CN=C1
138.126
138.13
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes