Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-phenoxybenzoate, 99%


Quantity 250g

Manufacturer: alfa aesar™  L1458430

 View more versions of this product

Catalog No. AAL1458430

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 228.25
Chemical Name or Material Methyl 2-phenoxybenzoate
Assay 99%
Molecular Formula C14H12O3
MDL Number MFCD03093064
CAS 21905-56-6
Synonym 2-Phenoxybenzoic acid methyl ester
Beilstein 2115719
Boiling Point 130°C (0.5mmHg)
Quantity 250g

Specifications

Chemical Name or Material Methyl 2-phenoxybenzoate
CAS 21905-56-6
Molecular Formula C14H12O3
Formula Weight 228.25
MDL Number MFCD03093064
Beilstein 2115719
Synonym 2-Phenoxybenzoic acid methyl ester