missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl-2-pentanone, 98.5% min., ACS, MilliporeSigma™
Supplier: Technidata MX13003
Specifications
Methyl-2-pentanone | |
108-10-1 | |
C6H12O | |
4 L | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
4-methylpentan-2-one | |
7909 | |
1.16 | |
Liquid |
For ACS analysis | |
98.5%, min. | |
(CH3)2CHCH2COCH3 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
CC(C)CC(=O)C | |
100.161 | |
CHEBI:82344 | |
ACS/NF/USP |
Chemical Identifiers
108-10-1 | |
100.161 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
CHEBI:82344 | |
CC(C)CC(=O)C |
C6H12O | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
7909 | |
4-methylpentan-2-one |
Safety and Handling
Recommended Storage : Room Temperature