missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2-oxo-2H-pyran-3-carboxylate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2593206
Description
Methyl 2-oxo-2H-pyran-3-carboxylate is used as a reagent to investigate alfa, beta-unsaturated carbonyl compounds for their cytotoxic profiles against oral human normal and tumor cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 2-oxo-2H-pyran-3-carboxylate is used as a reagent to investigate alfa, beta-unsaturated carbonyl compounds for their cytotoxic profiles against oral human normal and tumor cells.
Notes
Incompatible with strong oxidizing agents.
Specifications
Methyl 2-oxo-2H-pyran-3-carboxylate | |
74°C to 77°C | |
C7H6O4 | |
5 g | |
methyl 2-oxo-2h-pyran-3-carboxylate, 3-carbomethoxy-2-pyrone, methyl 2-pyrone-3-carboxylate, 2-oxo-2h-pyran-3-carboxylic acid methyl ester, 2h-pyran-3-carboxylic acid, 2-oxo-, methyl ester, methoxycarbonyl pyranone, acmc-1clg5, ksc496k6b | |
COC(=O)C1=CC=COC1=O | |
154.121 | |
154.12 |
25991-27-9 | |
146°C to 148°C (0.75 mmHg) | |
MFCD00075443 | |
1424749 | |
GJVZWOMUTYNUCE-UHFFFAOYSA-N | |
methyl 2-oxopyran-3-carboxylate | |
117704 | |
98% |
Chemical Identifiers
25991-27-9 | |
154.121 | |
GJVZWOMUTYNUCE-UHFFFAOYSA-N | |
117704 | |
COC(=O)C1=CC=COC1=O |
C7H6O4 | |
MFCD00075443 | |
methyl 2-oxo-2h-pyran-3-carboxylate, 3-carbomethoxy-2-pyrone, methyl 2-pyrone-3-carboxylate, 2-oxo-2h-pyran-3-carboxylic acid methyl ester, 2h-pyran-3-carboxylic acid, 2-oxo-, methyl ester, methoxycarbonyl pyranone, acmc-1clg5, ksc496k6b | |
methyl 2-oxopyran-3-carboxylate |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 247-397-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only