missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-(N-phthalimidooxy)-2-methylpropionate, 98%

Manufacturer:  Alfa Aesar™ H6113103

 View more versions of this product

Catalog No. AAH6113103



Specifications

Specifications

Methyl 2-(N-phthalimidooxy)-2-methylpropionate
84209-07-4
277.27
98%
1g
Methyl 2-(1,3-dioxo-1,3-dihydro-2-isoindolyloxy)-2-methylpropionate
Solid

Chemical Identifiers

84209-07-4
Methyl 2-(1,3-dioxo-1,3-dihydro-2-isoindolyloxy)-2-methylpropionate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents