Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-methylvalerate, 98%

Chemical Identifiers

Chemical Name or Material Methyl 2-methylvalerate
CAS 2177-77-7
Molecular Formula CH3CH2CH2CH(CH3)CO2CH3
Formula Weight 130.19g/mol
MDL Number MFCD00036653
UN Number UN3272
Synonym Methyl 2-methylpentanoate; 2-Methylvaleric acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1981214

 
alfa aesar™
A19812-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1981222

 
alfa aesar™
A19812-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 85°C (18mmHg)
Quantity 25g
Flash Point 35°C (95°F)
Chemical Name or Material Methyl 2-methylvalerate
CAS 2177-77-7
Assay 98%
Molecular Formula CH3CH2CH2CH(CH3)CO2CH3
Formula Weight 130.19g/mol
MDL Number MFCD00036653
UN Number UN3272
Synonym Methyl 2-methylpentanoate; 2-Methylvaleric acid methyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.