Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-(methylthio)propionate, 99%

$76.39 - $269.72

Chemical Identifiers

Chemical Name or Material Methyl 2-(methylthio)propionate
CAS 61366-76-5
Molecular Formula C5H10O2S
Formula Weight 134.2
MDL Number MFCD02683079
Synonym Methyl 2-(methylmercapto)propionate; 2-(Methylthio)propionic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2197906

  SDS
alfa aesar™
B2197906
5g Each for $76.39

AAB2197914

  SDS
alfa aesar™
B2197914
25g Each for $269.72
Description & Specifications

Specifications

Formula Weight 134.2
CAS 61366-76-5
Synonym Methyl 2-(methylmercapto)propionate; 2-(Methylthio)propionic acid methyl ester
Quantity 5g
Assay 99%
MDL Number MFCD02683079
Chemical Name or Material Methyl 2-(methylthio)propionate
Molecular Formula C5H10O2S

Applications
Methyl 2-(methylthio)propionate is used in the contribution of overall aroma perception of the ripe fruit.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.