Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-(methylthio)butyrate, 98%

$68.50 - $68.50

Chemical Identifiers

Chemical Name or Material Methyl 2-(methylthio)butyrate
CAS 51534-66-8
Molecular Formula C6H12O2S
Formula Weight 148.23
MDL Number MFCD00013393
Beilstein 1925141
Synonym 2-(Methylthio)butyric acid methyl ester; Methyl 2-(methylmercapto)butyrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2230306

  SDS
alfa aesar™
B2230306
5g Each for $68.50
Description & Specifications

Specifications

Formula Weight 148.23
Chemical Name or Material Methyl 2-(methylthio)butyrate
Assay 98%
Molecular Formula C6H12O2S
MDL Number MFCD00013393
CAS 51534-66-8
Beilstein 1925141
Synonym 2-(Methylthio)butyric acid methyl ester; Methyl 2-(methylmercapto)butyrate
Health Hazard 1 Warning
Boiling Point 103° to 104°C (35mmHg)
Quantity 5g
Flash Point 64°C (147°F)
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.