missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-methyl-3-furoate, 97%

Manufacturer:  Alfa Aesar™ A1197914

 View more versions of this product

Catalog No. AAA1197914



Specifications

Specifications

Methyl 2-methyl-3-furoate
71°C to 72°C (20mmHg)
6141-58-8
MFCD00003249
methyl 2-methyl-3-furoate, methyl 2-methyl-3-furancarboxylate, 2-methyl-furan-3-carboxylic acid methyl ester, 3-furancarboxylic acid, 2-methyl-, methyl ester, 3-furoic acid, 2-methyl-, methyl ester, methyl2-methyl-3-furoate, acmc-1bcvo, methyl 2-methyl-3-furoate #, methyl-2-methyl-3-furoate, 2-methylfuran-3-carboxylic acid methyl
CC1=C(C=CO1)C(=O)OC
140.138
140.14
63°C (145°F)
25g
C7H8O3
1342040
UVRRIABXNIGUJZ-UHFFFAOYSA-N
methyl 2-methylfuran-3-carboxylate
80237
97%

Chemical Identifiers

6141-58-8
140.138
UVRRIABXNIGUJZ-UHFFFAOYSA-N
80237
CC1=C(C=CO1)C(=O)OC
C7H8O3
MFCD00003249
methyl 2-methyl-3-furoate, methyl 2-methyl-3-furancarboxylate, 2-methyl-furan-3-carboxylic acid methyl ester, 3-furancarboxylic acid, 2-methyl-, methyl ester, 3-furoic acid, 2-methyl-, methyl ester, methyl2-methyl-3-furoate, acmc-1bcvo, methyl 2-methyl-3-furoate #, methyl-2-methyl-3-furoate, 2-methylfuran-3-carboxylic acid methyl
methyl 2-methylfuran-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents