Please login to your online account to display your discounted pricing

Methyl 2-methyl-3-furancarboxylate, 97%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  208975000

 View more versions of this product

Catalog No. AC208975000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 140.14
Quantity 500g
Color Clear yellow to brown
EINECS Number 228-132-6
Density 1.1100g/mL
Refractive Index 1.4720 to 1.4780 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
MDL Number MFCD00003249
Assay Percent Range 97%
Chemical Name or Material Methyl 2-methyl-3-furancarboxylate
Molecular Formula C7 H8 O3
CAS 6141-58-8

Specifications

Chemical Name or Material Methyl 2-methyl-3-furancarboxylate
Name Note 97%
CAS 6141-58-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7 H8 O3
Formula Weight 140.14
MDL Number MFCD00003249