Please login to your online account to display your discounted pricing

Methyl 2-methyl-3-furancarboxylate, 97%, ACROS Organics™

$411.76 - $411.76

Chemical Identifiers

Chemical Name or Material Methyl 2-methyl-3-furancarboxylate
Name Note 97%
CAS 6141-58-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7 H8 O3
Formula Weight 140.14
MDL Number MFCD00003249
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC208975000

  SDS
thermo fisher scientific
208975000
Glass bottle 500g Each for $411.76
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 140.14
Quantity 500g
Color Clear yellow to brown
EINECS Number 228-132-6
Density 1.1100g/mL
Refractive Index 1.4720 to 1.4780 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
MDL Number MFCD00003249
Assay Percent Range 97%
Chemical Name or Material Methyl 2-methyl-3-furancarboxylate
Molecular Formula C7 H8 O3
CAS 6141-58-8