missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2-methoxybenzoate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1770314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 2-methoxybenzoate | |
1.16 | |
>110°C (230°F) | |
C9H10O3 | |
MFCD00008423 | |
2207356 | |
PFYHAAAQPNMZHO-UHFFFAOYSA-N | |
methyl 2-methoxybenzoate | |
61151 | |
≥98% |
606-45-1 | |
246°C to 248°C | |
Characteristic | |
1.534 | |
25 g | |
methyl o-anisate, dimethyl salicylate, o-anisic acid, methyl ester, methyl o-methoxybenzoate, o-methoxy methyl benzoate, o-methoxybenzoic acid methyl ester, methylsalicylate methyl ether, 2-methoxybenzoic acid methyl ester, benzoic acid, 2-methoxy-, methyl ester, 2-methoxybenzoic acid, methyl ester | |
COC1=CC=CC=C1C(=O)OC | |
166.176 | |
166.18 |
Chemical Identifiers
606-45-1 | |
166.176 | |
PFYHAAAQPNMZHO-UHFFFAOYSA-N | |
61151 | |
COC1=CC=CC=C1C(=O)OC |
C9H10O3 | |
MFCD00008423 | |
methyl o-anisate, dimethyl salicylate, o-anisic acid, methyl ester, methyl o-methoxybenzoate, o-methoxy methyl benzoate, o-methoxybenzoic acid methyl ester, methylsalicylate methyl ether, 2-methoxybenzoic acid methyl ester, benzoic acid, 2-methoxy-, methyl ester, 2-methoxybenzoic acid, methyl ester | |
methyl 2-methoxybenzoate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-118-6
RTECSNumber : DH3549000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only