missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2-methoxy-5-nitrobenzoate, 98%, Thermo Scientific Chemicals
$170.44 - $596.55
Chemical Identifiers
CAS | 34841-11-7 |
---|---|
Molecular Formula | C9H9NO5 |
Molecular Weight (g/mol) | 211.173 |
MDL Number | MFCD00673023 |
InChI Key | DOBFJVVTBNTGCW-UHFFFAOYSA-N |
Synonym | 2-methoxy-5-nitrobenzoic acid methyl ester, methyl2-methoxy-5-nitrobenzoate, methyl-2-methoxy-5-nitrobenzoate, 5-nitro-o-anisic acid methyl ester, pubchem14052, acmc-1adrt, methyl 5-nitro-o-anisate, asischem d13383, rarechem al bf 0877, timtec-bb sbb014757 |
PubChem CID | 7020925 |
IUPAC Name | methyl 2-methoxy-5-nitrobenzoate |
SMILES | COC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2186306
|
Thermo Scientific Chemicals
B2186306 |
5 g |
Each for $170.44
|
|
|||||
AAB2186314
|
Thermo Scientific Chemicals
B2186314 |
25 g |
Each for $596.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34841-11-7 | |
211.173 | |
DOBFJVVTBNTGCW-UHFFFAOYSA-N | |
7020925 | |
COC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC |
C9H9NO5 | |
MFCD00673023 | |
2-methoxy-5-nitrobenzoic acid methyl ester, methyl2-methoxy-5-nitrobenzoate, methyl-2-methoxy-5-nitrobenzoate, 5-nitro-o-anisic acid methyl ester, pubchem14052, acmc-1adrt, methyl 5-nitro-o-anisate, asischem d13383, rarechem al bf 0877, timtec-bb sbb014757 | |
methyl 2-methoxy-5-nitrobenzoate |
Specifications
34841-11-7 | |
C9H9NO5 | |
5 g | |
DOBFJVVTBNTGCW-UHFFFAOYSA-N | |
methyl 2-methoxy-5-nitrobenzoate | |
7020925 | |
98% |
99°C to 102°C | |
MFCD00673023 | |
2-methoxy-5-nitrobenzoic acid methyl ester, methyl2-methoxy-5-nitrobenzoate, methyl-2-methoxy-5-nitrobenzoate, 5-nitro-o-anisic acid methyl ester, pubchem14052, acmc-1adrt, methyl 5-nitro-o-anisate, asischem d13383, rarechem al bf 0877, timtec-bb sbb014757 | |
COC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)OC | |
211.173 | |
211.18 | |
Methyl 2-methoxy-5-nitrobenzoate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only