missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl-2-isocyanatobenzoate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440150050

 View more versions of this product

Catalog No. AC440150050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl-2-isocyanatobenzoate
99.0°C (0.5 Torr)
47.0°C to 50.0°C
1793-07-3
C9H7NO3
ATFKBWNEVZUSKC-UHFFFAOYSA-N
methyl 2-isocyanatobenzoate
519494
Solid or Crystalline Solid
Authentic
White
5g
96% min. (GC)
#, 2-carbomethoxyphenyl isocyanate, 2-carbomethoxyphenylisocyanate, 2-methoxycarbonyl phenyl isocyanate, 2-methoxycarbonyl phenylisocyanate, 2-methoxycarbonylphenyl isocyanate, acmc-1bqmd, benzoic acid, 2-isocyanato-, methyl ester, methyl 2 isocyanatobenzoate, methyl-2-isocyanatobenzoate
COC(=O)C1=CC=CC=C1N=C=O
177.16
177.16
97%

Chemical Identifiers

1793-07-3
177.16
#, 2-carbomethoxyphenyl isocyanate, 2-carbomethoxyphenylisocyanate, 2-methoxycarbonyl phenyl isocyanate, 2-methoxycarbonyl phenylisocyanate, 2-methoxycarbonylphenyl isocyanate, acmc-1bqmd, benzoic acid, 2-isocyanato-, methyl ester, methyl 2 isocyanatobenzoate, methyl-2-isocyanatobenzoate
methyl 2-isocyanatobenzoate
C9H7NO3
ATFKBWNEVZUSKC-UHFFFAOYSA-N
519494
COC(=O)C1=CC=CC=C1N=C=O
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic if inhaled.
May cause allergy or asthma symptoms

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

GHS Signal Word: Danger

SDS Documents