missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-iodobenzoate, 99+%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC249831000

 View more versions of this product

Catalog No. AC249831000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl 2-iodobenzoate
1.7300g/mL
272.0°C to 274.0°C
Authentic
IC6H4COOCH3
Glass bottle
2-iodo-benzoic acid methyl ester, 2-iodobenzoic acid methyl ester, acmc-209mp0, benzoic acid, 2-iodo-, methyl ester, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, methyl o-iodobenzoate, methyl-2-iodobenzoate, methyl2-iodobenzoate, pubchem10568
Solubility in water: insoluble. Other solubilities: soluble in polar solvents,insoluble in non-polar solvents
1.73
methyl 2-iodobenzoate
69132
Crystalling or Flaky Powder
610-97-9
White
99% min. (GC)
C8H7IO2
MFCD00016351
1.6020 to 1.6040
100mL
BXXLTVBTDZXPTN-UHFFFAOYSA-N
COC(=O)C1=CC=CC=C1I
262.04
262.04
99+%

Chemical Identifiers

610-97-9
262.04
BXXLTVBTDZXPTN-UHFFFAOYSA-N
69132
COC(=O)C1=CC=CC=C1I
C8H7IO2
MFCD00016351
2-iodo-benzoic acid methyl ester, 2-iodobenzoic acid methyl ester, acmc-209mp0, benzoic acid, 2-iodo-, methyl ester, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, methyl o-iodobenzoate, methyl-2-iodobenzoate, methyl2-iodobenzoate, pubchem10568
methyl 2-iodobenzoate
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-243-6

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only