missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-iodobenzoate, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 249830250

 View more versions of this product

Catalog No. AC249830250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Methyl 2-iodobenzoate
Authentic
1.6020 to 1.6040
272.0°C to 274.0°C
25mL
99% min. (GC)
IC6H4COOCH3
2-iodo-benzoic acid methyl ester, 2-iodobenzoic acid methyl ester, acmc-209mp0, benzoic acid, 2-iodo-, methyl ester, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, methyl o-iodobenzoate, methyl-2-iodobenzoate, methyl2-iodobenzoate, pubchem10568
BXXLTVBTDZXPTN-UHFFFAOYSA-N
methyl 2-iodobenzoate
69132
Crystalling or Flaky Powder
1.7300g/mL
Glass bottle
1.73
White
610-97-9
C8H7IO2
MFCD00016351
Solubility in water: insoluble. Other solubilities: soluble in polar solvents,insoluble in non-polar solvents
COC(=O)C1=CC=CC=C1I
262.04
262.04
99+%

Chemical Identifiers

610-97-9
262.04
BXXLTVBTDZXPTN-UHFFFAOYSA-N
69132
COC(=O)C1=CC=CC=C1I
C8H7IO2
MFCD00016351
2-iodo-benzoic acid methyl ester, 2-iodobenzoic acid methyl ester, acmc-209mp0, benzoic acid, 2-iodo-, methyl ester, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, methyl o-iodobenzoate, methyl-2-iodobenzoate, methyl2-iodobenzoate, pubchem10568
methyl 2-iodobenzoate
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-243-6

SDS Documents