Please login to your online account to display your discounted pricing

Methyl 2-hydroxyisobutyrate 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  126970050

 View more versions of this product

Catalog No. AC126970050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 118.13
Assay ≥98.5%
Quantity 5g
Color Clear colorless
EINECS Number 218-301-2
TSCA TSCA
Density 1.03g/mL
Beilstein 03, II, 223
Packaging Glass bottle
Refractive Index 1.4100 to 1.4120 (20°C, 589nm)
Synonym Methyl 2-methyllactate
Chemical Name or Material Methyl 2-hydroxyisobutyrate
Name Note 99%
Assay Percent Range 99%
Molecular Formula C5H10O3
Linear Formula (CH3)2C(OH)CO2CH3
MDL Number MFCD00004457
CAS 2110-78-3
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Methyl 2-hydroxyisobutyrate
Name Note 99%
CAS 2110-78-3
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H10O3
Linear Formula (CH3)2C(OH)CO2CH3
Formula Weight 118.13
MDL Number MFCD00004457
Synonym Methyl 2-methyllactate
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents