Please login to your online account to display your discounted pricing

Methyl 2-hexynoate, +98%, ACROS Organics™

$63.40 - $63.40

Chemical Identifiers

Chemical Name or Material Methyl 2-hexynoate
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H10O2
Linear Formula CH3(CH2)2C2CCOOCH3
Formula Weight 126.15
MDL Number MFCD00041642
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382090050

  SDS
thermo fisher scientific
AC382090050
Glass bottle 5g Each for $63.40

AC382090250

  SDS
thermo fisher scientific
AC382090250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Liquid
Formula Weight 126.15
Quantity 5g
Color Clear colorless to yellow
Density 0.943g/mL
Molecular Formula C7H10O2
Linear Formula CH3(CH2)2C2CCOOCH3
CAS 18937-79-6
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl 2-hexynoate
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Assay Percent Range 98+%
MDL Number MFCD00041642
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.