Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-hexynoate, 98+%

$75.11 - $75.11

Chemical Identifiers

Chemical Name or Material Methyl 2-hexynoate
CAS 18937-79-6
Molecular Formula CH3CH2CH2C≡CCO2CH3
Formula Weight 126.16
MDL Number MFCD00041642
Beilstein 1750018
Synonym 2-Hexynoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2306506

 
alfa aesar™
B2306506
5g Each for $75.11
Description & Specifications

Specifications

Formula Weight 126.16
Chemical Name or Material Methyl 2-hexynoate
Assay ≥98%
Molecular Formula CH3CH2CH2C≡CCO2CH3
MDL Number MFCD00041642
CAS 18937-79-6
Beilstein 1750018
Synonym 2-Hexynoic acid methyl ester
Health Hazard 1 Warning
Recommended Storage 2° to 8°C
Flash Point 67°C (152°F)
Boiling Point 64° to 66°C (10mmHg)
Quantity 5g
Safety and Handling

Warning

2° to 8°C