missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-furoate 98%, ACROS Organics™

Manufacturer:  Acros Organics 156871000

 View more versions of this product

Catalog No. AC156871000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl 2-furoate
73°C
Glass bottle
1.176
Yellow to Brown
98%
98%
MFCD00003236
HDJLSECJEQSPKW-UHFFFAOYSA-N
methyl furan-2-carboxylate
11902
Liquid
1.1760g/mL
Authentic
1.4830 to 1.4890
181.0°C
100g
611-13-2
C6H6O3
methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester
COC(=O)C1=CC=CO1
126.111
126.11
98%

Chemical Identifiers

611-13-2
126.111
HDJLSECJEQSPKW-UHFFFAOYSA-N
11902
COC(=O)C1=CC=CO1
C6H6O3
MFCD00003236
methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester
methyl furan-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-254-6

RTECSNumber : LV1950000

TSCA : TSCA

SDS Documents