Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (2-furfurylthio)acetate, 97%

$450.53 - $450.53

Chemical Identifiers

Chemical Name or Material Methyl (2-furfurylthio)acetate
CAS 108499-33-8
Molecular Formula C8H10O2S
Formula Weight 186.23
MDL Number MFCD02283460
Synonym (2-Furfurylthio)acetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2271722

  SDS
alfa aesar™
B2271722
100g Each for $450.53

AAB2271706

  SDS
alfa aesar™
B2271706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2271714

  SDS
alfa aesar™
B2271714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Methyl (2-furfurylthio)acetate
CAS 108499-33-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C8H10O2S
Formula Weight 186.23
MDL Number MFCD02283460
Synonym (2-Furfurylthio)acetic acid methyl ester
Boiling Point 95°C (3mmHg)
Quantity 100g
Flash Point >100°C (212°F)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.