Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-formylthiophene-4-carboxylate, 95%

$83.93 - $622.30

Chemical Identifiers

Chemical Name or Material Methyl 2-formylthiophene-4-carboxylate
CAS 67808-66-6
Molecular Formula C7H6O3S
Formula Weight 170.19
MDL Number MFCD01859945
Synonym 2-Formylthiophene-4-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2759703

  SDS
alfa aesar™
H2759703
1g Each for $221.50

AAH2759706

  SDS
alfa aesar™
H2759706
5g Each for $622.30

AAH27597MD

  SDS
alfa aesar™
H27597MD
250mg Each for $83.93
Description & Specifications

Specifications

Formula Weight 170.19
Quantity 1g
Melting Point 113° to 117°C
Molecular Formula C7H6O3S
MDL Number MFCD01859945
Health Hazard 1 Warning
CAS 67808-66-6
Chemical Name or Material Methyl 2-formylthiophene-4-carboxylate
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 2-Formylthiophene-4-carboxylic acid methyl ester
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.