Please login to your online account to display your discounted pricing

Methyl 2-fluorophenylacetate, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC304641000

 View more versions of this product

Catalog No. AC304641000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5% (GC)
Formula Weight 168.06
Quantity 100g
Color Clear colorless
Chemical Name or Material Methyl 2-fluorophenylacetate
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
Assay Percent Range 99%
CAS 57486-67-6
Molecular Formula C9H9FO2

Specifications

Chemical Name or Material Methyl 2-fluorophenylacetate
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C9H9FO2
Formula Weight 168.06