Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-ethylacetoacetate, 97%

$76.05 - $238.59

Chemical Identifiers

Chemical Name or Material Methyl 2-ethylacetoacetate
CAS 51756-08-2
Molecular Formula C7H12O3
Formula Weight 144.17
MDL Number MFCD01632136
Synonym 2-Ethylacetoacetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2136814

 
alfa aesar™
B2136814
25g Each for $76.05

AAB2136822

 
alfa aesar™
B2136822
100g Each for $238.59
Description & Specifications

Specifications

Formula Weight 144.17
Chemical Name or Material Methyl 2-ethylacetoacetate
TSCA TSCA
Assay 97%
Molecular Formula C7H12O3
MDL Number MFCD01632136
CAS 51756-08-2
Synonym 2-Ethylacetoacetic acid methyl ester
Health Hazard 1 Warning
Boiling Point 189° to 191°C
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.