Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-diphenylphosphino-1-naphthoate, 98%

$81.58 - $1,103.66

Chemical Identifiers

Chemical Name or Material Methyl 2-diphenylphosphino-1-naphthoate
CAS 178176-78-8
Molecular Formula C24H19O2P
Formula Weight 370.39
MDL Number MFCD12546191
Synonym 2-Diphenylphosphino-1-naphthoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3092803

 
alfa aesar™
H3092803
1g Each for $81.58

AAH3092806

 
alfa aesar™
H3092806
5g Each for $283.23

AAH3092814

 
alfa aesar™
H3092814
25g Each for $1,103.66
Description & Specifications

Specifications

Formula Weight 370.39
Chemical Name or Material Methyl 2-diphenylphosphino-1-naphthoate
Assay 98%
Molecular Formula C24H19O2P
MDL Number MFCD12546191
CAS 178176-78-8
Synonym 2-Diphenylphosphino-1-naphthoic acid methyl ester
Health Hazard 1 Warning
Melting Point 107° to 108°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.