Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-dimethylaminobenzoate, 95%

$64.34 - $233.55

Chemical Identifiers

Chemical Name or Material Methyl 2-dimethylaminobenzoate
CAS 10072-05-6
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00025858
Beilstein 2804944
Synonym 2-Dimethylaminobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1206604

  SDS
alfa aesar™
L1206604
2g Each for $64.34

AAL1206609

  SDS
alfa aesar™
L1206609
10g Each for $233.55
Description & Specifications

Specifications

Formula Weight 179.22
Chemical Name or Material Methyl 2-dimethylaminobenzoate
CAS 10072-05-6
Molecular Formula C10H13NO2
Synonym 2-Dimethylaminobenzoic acid methyl ester
Assay 95%
Flash Point 122°C (251°F)
Beilstein 2804944
Boiling Point 133° to 134°C (16mmHg)
Quantity 2g
MDL Number MFCD00025858