Alfa Aesar™ Methyl 2-dimethylaminobenzoate, 95%
Manufacturer:  Alfa Aesar™ L1206609
 View more versions of this product
Catalog No. AAL1206609
Specifications
Specifications
Methyl 2-dimethylaminobenzoate
133°C to 134°C (16mmHg)
10072-05-6
MFCD00025858
methyl 2-dimethylamino benzoate, methyl n,n-dimethylanthranilate, unii-77lqp0681t, n,n-dimethylanthranilic acid methyl ester, benzoic acid, 2-dimethylamino-, methyl ester, acmc-1bs8u, anthranilic acid, n,n-dimethyl-, methyl ester, methyl dimethyl anthranilate, methyln,n-dimethylanthranilate, methyl 2-dimethylamino-benzoate
CN(C)C1=CC=CC=C1C(=O)OC
179.219
179.22
122°C (251°F)
10g
C10H13NO2
2804944
ZCNSBHAIPOWHJE-UHFFFAOYSA-N
methyl 2-(dimethylamino)benzoate
82336
95%
Chemical Identifiers
10072-05-6
179.219
ZCNSBHAIPOWHJE-UHFFFAOYSA-N
82336
CN(C)C1=CC=CC=C1C(=O)OC
C10H13NO2
MFCD00025858
methyl 2-dimethylamino benzoate, methyl n,n-dimethylanthranilate, unii-77lqp0681t, n,n-dimethylanthranilic acid methyl ester, benzoic acid, 2-dimethylamino-, methyl ester, acmc-1bs8u, anthranilic acid, n,n-dimethyl-, methyl ester, methyl dimethyl anthranilate, methyln,n-dimethylanthranilate, methyl 2-dimethylamino-benzoate
methyl 2-(dimethylamino)benzoate
Documents