Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-cyanoisonicotinate, 97%

Chemical Identifiers

Chemical Name or Material Methyl 2-cyanoisonicotinate
CAS 94413-64-6
Formula Weight 162.15
MDL Number MFCD07367894
UN Number UN3439
Synonym Methyl 2-cyanopyridine-4-carboxylate; 2-Cyanopyridine-4-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6309003

  SDS
alfa aesar™
H6309003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6309006

  SDS
alfa aesar™
H6309006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63090MD

  SDS
alfa aesar™
H63090MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.15
Chemical Name or Material Methyl 2-cyanoisonicotinate
Physical Form Solid
Assay 97%
MDL Number MFCD07367894
CAS 94413-64-6
UN Number UN3439
Synonym Methyl 2-cyanopyridine-4-carboxylate; 2-Cyanopyridine-4-carboxylic acid methyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Color Pale brown
Quantity 1g

Applications
Methyl 2-cyanoisonicotinate can be used in the synthesis of various pharmaceutical and biologically active compounds, such as tetra substituted imidazolines as potent and selective neuropeptide Y (NPY) Y5 receptor antagonists.

Solubility
Slightly Soluble in water (6.2 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.