Learn More
Methyl 2-cyanoisonicotinate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6309006
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsMethyl 2-cyanoisonicotinate can be used in the synthesis of various pharmaceutical and biologically active compounds, such as tetra substituted imidazolines as potent and selective neuropeptide Y (NPY) Y5 receptor antagonists.
Solubility
Slightly Soluble in water (6.2 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Methyl 2-cyanoisonicotinate | |
Brown | |
MFCD07367894 | |
UN3439 | |
Slightly Soluble in water (6.2g/L) (25°C). | |
COC(=O)C1=CC(=NC=C1)C#N | |
162.148 | |
162.15 | |
Solid |
94413-64-6 | |
C8H6N2O2 | |
5 g | |
methyl 2-cyanoisonicotinate, 2-cyano-4-pyridine carboxylic acid methyl ester, 4-pyridinecarboxylicacid, 2-cyano-, methyl ester, 4-pyridinecarboxylic acid, 2-cyano-, methyl ester, methyl 2-cyanois, methyl 2-cyanoisonicotite, 2-cyano-4-carbomethoxypyridine, 2-cyanoisonicotinic acid methyl ester, 2-cyano-isonicotinic acid methyl ester, 2-cyano-4-pyridine-carboxylic acid methyl ester | |
ORVHMLCJEKDDAX-UHFFFAOYSA-N | |
methyl 2-cyanopyridine-4-carboxylate | |
12132796 | |
97% |
Chemical Identifiers
94413-64-6 | |
162.148 | |
ORVHMLCJEKDDAX-UHFFFAOYSA-N | |
12132796 | |
COC(=O)C1=CC(=NC=C1)C#N |
C8H6N2O2 | |
MFCD07367894 | |
methyl 2-cyanoisonicotinate, 2-cyano-4-pyridine carboxylic acid methyl ester, 4-pyridinecarboxylicacid, 2-cyano-, methyl ester, 4-pyridinecarboxylic acid, 2-cyano-, methyl ester, methyl 2-cyanois, methyl 2-cyanoisonicotite, 2-cyano-4-carbomethoxypyridine, 2-cyanoisonicotinic acid methyl ester, 2-cyano-isonicotinic acid methyl ester, 2-cyano-4-pyridine-carboxylic acid methyl ester | |
methyl 2-cyanopyridine-4-carboxylate |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures