Please login to your online account to display your discounted pricing

Methyl 2-(chlorosulfonyl)benzoate, 90%, Tech., ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  224160250

 View more versions of this product

Catalog No. AC224160250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥88% (ex Cl)
Physical Form Crystalline Powder or Chunks
Formula Weight 234.66
Quantity 25g
Color White to beige or pink
TSCA TSCA
Melting Point 58° to 64°C
Beilstein 11, 373
Chemical Name or Material Methyl 2-(chlorosulfonyl)benzoate
Name Note tech., 90%
Assay Percent Range 90%
Molecular Formula C8H7ClO4S
Linear Formula ClSO2C6H4COOCH3
MDL Number MFCD00009797
Synonym CBC
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
CAS 26638-43-7

Specifications

Chemical Name or Material Methyl 2-(chlorosulfonyl)benzoate
Name Note tech., 90%
CAS 26638-43-7
Assay ≥88% (ex Cl)
Assay Percent Range 90%
Molecular Formula C8H7ClO4S
Linear Formula ClSO2C6H4COOCH3
Formula Weight 234.66
MDL Number MFCD00009797
Synonym CBC
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents