missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-(chlorosulfonyl)benzoate, 90%, Tech., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 224160050

 View more versions of this product

Catalog No. AC224160050


 

Specifications Description & Specifications

Specifications

≥88% (ex Cl)
234.66
White to beige or pink
58° to 64°C
MFCD00009797
90%
C8H7ClO4S
tech., 90%
ClSO2C6H4COOCH3
Crystalline Powder or Chunks
5g
11, 373
Glass bottle
CBC
Authentic
26638-43-7
Methyl 2-(chlorosulfonyl)benzoate

Chemical Identifiers

26638-43-7
C8H7ClO4S
234.66
CBC
≥88% (ex Cl)
90%
ClSO2C6H4COOCH3
MFCD00009797
Methyl 2-(chlorosulfonyl)benzoate, tech., 90%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA