missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-(chlorosulfonyl)benzoate, 90%, Tech., ACROS Organics™

$66.31 - $234.75

Chemical Identifiers

CAS 26638-43-7
Assay Percent Range 90%
Molecular Formula C8H7ClO4S
Linear Formula ClSO2C6H4COOCH3
Formula Weight 234.66
MDL Number MFCD00009797
Synonym CBC
Chemical Name or Material Methyl 2-(chlorosulfonyl)benzoate, tech., 90%
Assay ≥88% (ex Cl)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC224160250 SDS Thermo Fisher Scientific
224160250
Glass bottle 25g Each for $234.75
AC224160050 SDS Thermo Fisher Scientific
224160050
Glass bottle 5g Each for $66.31
Specifications Description & Specifications

Chemical Identifiers

26638-43-7
C8H7ClO4S
234.66
CBC
≥88% (ex Cl)
90%
ClSO2C6H4COOCH3
MFCD00009797
Methyl 2-(chlorosulfonyl)benzoate, tech., 90%

Specifications

White to beige or pink
25g
Authentic
Crystalline Powder or Chunks
C8H7ClO4S
234.66
11, 373
Methyl 2-(chlorosulfonyl)benzoate, tech., 90%
58° to 64°C
26638-43-7
Glass bottle
90%
ClSO2C6H4COOCH3
MFCD00009797
CBC
≥88% (ex Cl)

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA