Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-chloropyridine-3-carboxylate, 98%

Chemical Identifiers

Chemical Name or Material Methyl 2-chloropyridine-3-carboxylate
CAS 40134-18-7
Formula Weight 171.58
MDL Number MFCD01318629
Beilstein 122129
Synonym 2-Chloropyridine-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6479806

 
alfa aesar™
H6479806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6479814

 
alfa aesar™
H6479814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6479822

 
alfa aesar™
H6479822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.58
Chemical Name or Material Methyl 2-chloropyridine-3-carboxylate
Refractive Index 1.5370
Physical Form Liquid
Assay 98%
MDL Number MFCD01318629
CAS 40134-18-7
Beilstein 122129
Synonym 2-Chloropyridine-3-carboxylic acid methyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.314g/mL
Boiling Point 150°C (1mmHg)
Color Colorless
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.