Learn More
Methyl 2-chloropropionate, 97%, Thermo Scientific Chemicals
$140.96 - $470.33
Chemical Identifiers
CAS | 17639-93-9 |
---|---|
Molecular Formula | C4H7ClO2 |
Molecular Weight (g/mol) | 122.55 |
MDL Number | MFCD00000863 |
InChI Key | JLEJCNOTNLZCHQ-UHFFFAOYNA-N |
Synonym | methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6f |
PubChem CID | 28659 |
IUPAC Name | methyl 2-chloropropanoate |
SMILES | COC(=O)C(C)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1854130
|
Thermo Scientific Chemicals
A1854130 |
250 g |
Each for $140.96
|
|
|||||
AAA185410B
|
Thermo Scientific Chemicals
A185410B |
1000 g |
Each for $470.33
|
|
|||||
Description
Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.
Solubility
Insoluble in water
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
17639-93-9 | |
122.55 | |
JLEJCNOTNLZCHQ-UHFFFAOYNA-N | |
28659 | |
COC(=O)C(C)Cl |
C4H7ClO2 | |
MFCD00000863 | |
methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6f | |
methyl 2-chloropropanoate |
Specifications
17639-93-9 | |
1.14 | |
38°C (100°F) | |
1.417 | |
250 g | |
1421535 | |
Insoluble in water | |
COC(=O)C(C)Cl | |
122.55 | |
122.55 | |
Methyl 2-chloropropionate |
<-10°C | |
132°C to 133°C | |
C4H7ClO2 | |
MFCD00000863 | |
UN2933 | |
methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6f | |
JLEJCNOTNLZCHQ-UHFFFAOYNA-N | |
methyl 2-chloropropanoate | |
28659 | |
97% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYL 2-CHLOROPROPIONATE
EINECSNumber : 241-624-5
RTECSNumber : UE9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only