Please login to your online account to display your discounted pricing

Methyl 2-chlorophenylacetate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 2-chlorophenylacetate
Name Note 98%
CAS 57486-68-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9ClO2
Linear Formula ClC6H4CHCOCH3
Formula Weight 184.62
MDL Number MFCD00137445
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351040050

  SDS
fisher scientific acros
35104-0050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC351040250

  SDS
fisher scientific acros
35104-0250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 184.62
Quantity 5g
Color Clear colorless
Beilstein 09, 448
EINECS Number 260-764-8
Density 1.21g/mL
Chemical Name or Material Methyl 2-chlorophenylacetate
Refractive Index 1.5240 to 1.5260 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H9ClO2
Linear Formula ClC6H4CHCOCH3
MDL Number MFCD00137445
CAS 57486-68-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.