Learn More
Methyl 2-chlorophenylacetate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 351040250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl 2-chlorophenylacetate | |
57486-68-7 | |
Authentic | |
Glass bottle | |
1.5240 to 1.526 | |
MFCD00137445 | |
09, 448 | |
methyl 2-chlorophenylacetate, methyl 2-2-chlorophenyl acetate, methyl 2-chlorophenyl acetate, 2-chlorophenyl acetic acid methyl ester, benzeneacetic acid, 2-chloro-, methyl ester, 2-chlorophenylacetic acid methylester, 2-chlorobenzeneacetic acid methyl ester, acmc-209lyj, methyl2-chlorophenylacetate | |
COC(=O)CC1=CC=CC=C1Cl | |
184.62 | |
184.62 |
98% | |
1.2100g/mL | |
97.5% min. (GC) | |
C9H9ClO2 | |
ClC6H4CH2CO2CH3 | |
25g | |
1.21 | |
KHBWTRFWQROKJZ-UHFFFAOYSA-N | |
methyl 2-(2-chlorophenyl)acetate | |
93683 | |
98% |
Chemical Identifiers
57486-68-7 | |
184.62 | |
KHBWTRFWQROKJZ-UHFFFAOYSA-N | |
93683 | |
COC(=O)CC1=CC=CC=C1Cl |
C9H9ClO2 | |
MFCD00137445 | |
methyl 2-chlorophenylacetate, methyl 2-2-chlorophenyl acetate, methyl 2-chlorophenyl acetate, 2-chlorophenyl acetic acid methyl ester, benzeneacetic acid, 2-chloro-, methyl ester, 2-chlorophenylacetic acid methylester, 2-chlorobenzeneacetic acid methyl ester, acmc-209lyj, methyl2-chlorophenylacetate | |
methyl 2-(2-chlorophenyl)acetate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 260-764-8
RUO â Research Use Only