missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Chloronicotinate, TCI America™

$19.86 - $56.53

Chemical Identifiers

CAS 40134-18-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD01318629
InChI Key MYGAJZBZLONIBZ-UHFFFAOYSA-N
Synonym methyl 2-chloronicotinate, 2-chloronicotinic acid methyl ester, 2-chloro-3-pyridinecarboxylic acid methyl ester, methyl2-chloronicotinate, methyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, methyl ester, 2-chloro-nicotinic acid methyl ester, methyl chloronicotinate, pubchem15917, acmc-1am6z
PubChem CID 819931
IUPAC Name methyl 2-chloropyridine-3-carboxylate
SMILES COC(=O)C1=C(N=CC=C1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M216525G View Documents TCI America
M216525G
25g Each for $56.53
 
M21655G View Documents TCI America
M21655G
5g Each for $19.86
 
Specifications

Chemical Identifiers

40134-18-7
171.58
MYGAJZBZLONIBZ-UHFFFAOYSA-N
819931
COC(=O)C1=C(N=CC=C1)Cl
C7H6ClNO2
MFCD01318629
methyl 2-chloronicotinate, 2-chloronicotinic acid methyl ester, 2-chloro-3-pyridinecarboxylic acid methyl ester, methyl2-chloronicotinate, methyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, methyl ester, 2-chloro-nicotinic acid methyl ester, methyl chloronicotinate, pubchem15917, acmc-1am6z
methyl 2-chloropyridine-3-carboxylate

Specifications

70°C
25g
C7H6ClNO2
methyl 2-chloronicotinate, 2-chloronicotinic acid methyl ester, 2-chloro-3-pyridinecarboxylic acid methyl ester, methyl2-chloronicotinate, methyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, methyl ester, 2-chloro-nicotinic acid methyl ester, methyl chloronicotinate, pubchem15917, acmc-1am6z
COC(=O)C1=C(N=CC=C1)Cl
171.58
171.58
≥98.0% (GC)
Undesignated
40134-18-7
MFCD01318629
MYGAJZBZLONIBZ-UHFFFAOYSA-N
methyl 2-chloropyridine-3-carboxylate
819931
Clear Liquid at 20°C
Methyl 2-Chloronicotinate
Safety and Handling

Safety and Handling

TSCA : No