missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Chloronicotinate 98.0+%, TCI America™

Manufacturer:  Tci America M216525G

 View more versions of this product

Catalog No. M216525G



Specifications

Specifications

Methyl 2-Chloronicotinate
Undesignated
40134-18-7
MFCD01318629
MYGAJZBZLONIBZ-UHFFFAOYSA-N
methyl 2-chloropyridine-3-carboxylate
819931
Clear Liquid at 20°C
70°C
25g
C7H6ClNO2
2-chloro-3-pyridinecarboxylic acid methyl ester, 2-chloro-nicotinic acid methyl ester, 2-chloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-chloro-, methyl ester, acmc-1am6z, methyl 2-chloro-3-pyridinecarboxylate, methyl 2-chloronicotinate, methyl chloronicotinate, methyl2-chloronicotinate, pubchem15917
COC(=O)C1=C(N=CC=C1)Cl
171.58
171.58
≥98.0% (GC)

Chemical Identifiers

40134-18-7
171.58
MYGAJZBZLONIBZ-UHFFFAOYSA-N
819931
COC(=O)C1=C(N=CC=C1)Cl
C7H6ClNO2
MFCD01318629
2-chloro-3-pyridinecarboxylic acid methyl ester, 2-chloro-nicotinic acid methyl ester, 2-chloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-chloro-, methyl ester, acmc-1am6z, methyl 2-chloro-3-pyridinecarboxylate, methyl 2-chloronicotinate, methyl chloronicotinate, methyl2-chloronicotinate, pubchem15917
methyl 2-chloropyridine-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions