missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-chlorobenzoate, 98%

Manufacturer:  Alfa Aesar™ A1123736

 View more versions of this product

Catalog No. AAA1123736



Specifications

Specifications

Methyl 2-chlorobenzoate
98%
MFCD00016337
2-Chlorobenzoic acid methyl ester
COC(=O)C1=CC=CC=C1Cl
170.592
233°C to 235°C
500g
610-96-8
C8H7ClO2
1364684
JAVRNIFMYIJXIE-UHFFFAOYSA-N
methyl 2-chlorobenzoate
11895
170.6
108°C (226°F)

Chemical Identifiers

610-96-8
170.592
JAVRNIFMYIJXIE-UHFFFAOYSA-N
11895
COC(=O)C1=CC=CC=C1Cl
C8H7ClO2
MFCD00016337
2-Chlorobenzoic acid methyl ester
methyl 2-chlorobenzoate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents