missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Chloro-L-mandelate, TCI America™

$233.75 - $811.27

Chemical Identifiers

CAS 32345-60-1
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
InChI Key ZMPGBVQQIQSQED-QMMMGPOBSA-N
Synonym 2-Chloro-L-mandelic Acid Methyl Ester, Methyl (S)-2′C-Chloro-alpha-hydroxyphenylacetate, (S)-2′C-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester
PubChem CID 11183258
IUPAC Name methyl (2S)-2-(2-chlorophenyl)-2-hydroxyacetate
SMILES COC(=O)C(C1=CC=CC=C1Cl)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23831G View Documents TCI America
M23831G
1g Each for $233.75
 
M23835G View Documents TCI America
M23835G
5g Each for $811.27
 
Specifications

Chemical Identifiers

32345-60-1
200.618
2-Chloro-L-mandelic Acid Methyl Ester, Methyl (S)-2′C-Chloro-alpha-hydroxyphenylacetate, (S)-2′C-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester
methyl (2S)-2-(2-chlorophenyl)-2-hydroxyacetate
C9H9ClO3
ZMPGBVQQIQSQED-QMMMGPOBSA-N
11183258
COC(=O)C(C1=CC=CC=C1Cl)O

Specifications

Yellow
32345-60-1
2-Chloro-L-mandelic Acid Methyl Ester, Methyl (S)-2′C-Chloro-alpha-hydroxyphenylacetate, (S)-2′C-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester
COC(=O)C(C1=CC=CC=C1Cl)O
200.618
200.62
≥98.0% (GC)
1g
C9H9ClO3
ZMPGBVQQIQSQED-QMMMGPOBSA-N
methyl (2S)-2-(2-chlorophenyl)-2-hydroxyacetate
11183258
Clear Liquid at 20°C
Methyl 2-Chloro-L-mandelate
Safety and Handling

Safety and Handling

TSCA : No