missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2-Chloro-L-mandelate 98.0+%, TCI America™
Supplier: TCI America M23835G
Specifications
Methyl 2-Chloro-L-mandelate | |
Yellow | |
Clear Liquid at 20°C | |
2-Chloro-L-mandelic Acid Methyl Ester, Methyl (S)-2′-Chloro-alpha-hydroxyphenylacetate, (S)-2′-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester | |
COC(=O)C(C1=CC=CC=C1Cl)O | |
200.618 | |
200.62 |
32345-60-1 | |
5 g | |
C9H9ClO3 | |
ZMPGBVQQIQSQED-QMMMGPOBSA-N | |
methyl (2S)-2-(2-chlorophenyl)-2-hydroxyacetate | |
11183258 | |
≥98.0% (GC) |
Chemical Identifiers
32345-60-1 | |
200.618 | |
11183258 | |
COC(=O)C(C1=CC=CC=C1Cl)O |
C9H9ClO3 | |
ZMPGBVQQIQSQED-QMMMGPOBSA-N | |
methyl (2S)-2-(2-chlorophenyl)-2-hydroxyacetate |
Safety and Handling
TSCA : No