missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-chloro-6-methylpyridine-4-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5600603

 View more versions of this product

Catalog No. AAH5600603



Specifications

Specifications

Methyl 2-chloro-6-methylpyridine-4-carboxylate
58°C to 62°C
3998-90-1
MFCD04038391
BDWMGYZSQKGUFA-UHFFFAOYSA-N
methyl 2-chloro-6-methylpyridine-4-carboxylate
2759833
97%
124°C to 125°C (7mmHg)
1g
C8H8ClNO2
2-chloro-4-methoxycarbonyl-6-methylpyridine
CC1=NC(=CC(=C1)C(=O)OC)Cl
185.607
185.61

Chemical Identifiers

3998-90-1
185.607
BDWMGYZSQKGUFA-UHFFFAOYSA-N
2759833
CC1=NC(=CC(=C1)C(=O)OC)Cl
C8H8ClNO2
MFCD04038391
2-chloro-4-methoxycarbonyl-6-methylpyridine
methyl 2-chloro-6-methylpyridine-4-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c

H315-H317-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents